Safety boots, lace-up, Compositelite™ WORK Thor FC12
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FC12BKR39 FC12BKR40 FC12BKR41 FC12BKR42 FC12BKR43 FC12BKR44 FC12BKR45 FC12BKR46 FC12BKR38 FC12BKR48 FC12BKR47
PRTWFC12BKR39EA 84.1 EUR InStock
PRTWFC12BKR39 PRTWFC12BKR40 PRTWFC12BKR41 PRTWFC12BKR42 PRTWFC12BKR43 PRTWFC12BKR44 PRTWFC12BKR45 PRTWFC12BKR46 PRTWFC12BKR38 PRTWFC12BKR48 PRTWFC12BKR47
Safety boots, lace-up, Compositelite™ WORK Thor FC12
Schuhe und Stiefel
Metal-free, anti-static, lightweight boot made from water resistant nubuck fabric and a durable PU/rubber sole, resistant to oil and heat (200 °C).
  • Fur lining
  • Shock-absorbent heel
  • Composite toecap (200 J)
  • Puncture resistant composite midsole
Ideal for cold conditions.
Colour: Black
Zertifizierungen: EN 20345:2011+A1:2007; S3, HRO, SRC.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge