Safety shoes, lace-up, Compositelite™ ULTRA Vistula Trainer, FC67
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FC67BKR42 FC67BKR41 FC67BKR40 FC67BKR39 FC67BKR38 FC67BKR45 FC67BKR43 FC67BKR44 FC67BKR47 FC67BKR46
PRTWFC67BKR42EA 81.5 EUR InStock
PRTWFC67BKR42 PRTWFC67BKR41 PRTWFC67BKR40 PRTWFC67BKR39 PRTWFC67BKR38 PRTWFC67BKR45 PRTWFC67BKR43 PRTWFC67BKR44 PRTWFC67BKR47 PRTWFC67BKR46
Safety shoes, lace-up, Compositelite™ ULTRA Vistula Trainer, FC67
Schuhe und Stiefel
Metal-free, anti-static safety shoes made from micro PU upper plus a durable, dual-density phylon/rubber outer sole, resistant to oil and heat (300 °C).
  • Shock-absorbent heel
  • Composite toecap (200 J)
  • Puncture resistant, composite midsole
Suitable for light industrial use.
Colour: Black/blue
Zertifizierungen: EN 20345:2011 + A1:2007; S1P, SRC.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge