Safety boots, lace-up, Foyle, Portwest™ Pro, FD11
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FD11BKR42 FD11BKR38 FD11BKR39 FD11BKR40 FD11BKR41 FD11BKR43 FD11BKR44 FD11BKR45 FD11BKR46 FD11BKR47 FD11BKR48
PRTWFD11BKR42EA 90 EUR InStock
PRTWFD11BKR42 PRTWFD11BKR38 PRTWFD11BKR39 PRTWFD11BKR40 PRTWFD11BKR41 PRTWFD11BKR43 PRTWFD11BKR44 PRTWFD11BKR45 PRTWFD11BKR46 PRTWFD11BKR47 PRTWFD11BKR48
Safety boots, lace-up, Foyle, Portwest™ Pro, FD11
Schuhe und Stiefel
Anti-static safety boots made from full grain leather plus a dual density PU/rubber sole, resistant to heat (300 °C) are designed for tough outdoor working conditions and provides maximum protection.
  • Protective steel toecap
  • Steel midsole
  • Energy absorbing seat region
  • Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Fuel and oil resistant outsole
  • Wide fitting
Zertifizierungen: EN ISO 20345:2011 S3, SRC, HRO CI HI FO
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge