Compositelite™ S2 Safety Trainers, Slip-On, Low
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FD62WHR36 FD62WHR37 FD62WHR38 FD62WHR39 FD62WHR40 FD62WHR41 FD62WHR42 FD62WHR43 FD62WHR44 FD62WHR45 FD62WHR46 FD62WHR47 FD62WHR48
PRTWFD62WHR36EA 51.3 EUR InStock
PRTWFD62WHR36 PRTWFD62WHR37 PRTWFD62WHR38 PRTWFD62WHR39 PRTWFD62WHR40 PRTWFD62WHR41 PRTWFD62WHR42 PRTWFD62WHR43 PRTWFD62WHR44 PRTWFD62WHR45 PRTWFD62WHR46 PRTWFD62WHR47 PRTWFD62WHR48
Compositelite™ S2 Safety Trainers, Slip-On, Low
Schuhe und Stiefel
This metal-free mircofibre slip-on safety trainers offers an anti-microbial lining and a wipe clean upper. The slip-on silhouette aids an easy clean surface, reducing the possibility of contamination. Perfect for food, pharmaceutical, catering, and hospitality industries.
  • Padded collar for added comfort
  • Vegan friendly footwear
  • Composite toecap for added protection
  • Metal-free
  • Anti-static footwear
  • Energy absorbing seat region
  • Fuel- and oil-resistant outsole
  • SRC - Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Dual density sole unit
These slip-on safety trainers are anti-bacterial treated, with a perforated insole to help keep feet fresh and clean. The water-resistant upper prevents water penetration.
Zertifizierungen: EN ISO 20345: 2011 S2. CE certified.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge