Occupational boots, pull-on, non safety Wellington O4, FD90
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FD90GNR45 FD90GNR39 FD90GNR42 FD90GNR47 FD90GNR48 FD90GNR43 FD90GNR44 FD90GNR40 FD90GNR41 FD90GNR46
PRTWFD90GNR45EA 69.3 EUR InStock
PRTWFD90GNR45 PRTWFD90GNR39 PRTWFD90GNR42 PRTWFD90GNR47 PRTWFD90GNR48 PRTWFD90GNR43 PRTWFD90GNR44 PRTWFD90GNR40 PRTWFD90GNR41 PRTWFD90GNR46
Occupational boots, pull-on, non safety Wellington O4, FD90
Schuhe und Stiefel
Anti-static footwear with light weight and durable styles for all day comfort. Ideal for light work and leisure. Upper and sole are made from long lasting, breathable PU material keeps the feet healthy.
  • Energy absorbing seat region
  • 100% Waterproof keeping feet warm and dry
  • SRC - Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Fuel and oil resistant outsole
  • Cold insulation
  • 100% Metal free
Comfort footbed, lightweight PU construction for comfort and stability.
Colour: Green
Zertifizierungen: EN ISO 20347, CI, FO
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge