Steelite™ Tove Trainer S1P Safety Shoes, Lace-up, Low
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FT15BKB36 FT15BKB37 FT15BKB38 FT15BKB39 FT15BKB40 FT15BKB41 FT15BKB42 FT15BKB43 FT15BKB44 FT15BKB45 FT15BKB46 FT15BKB47 FT15BKB48
PRTWFT15BKB36EA 67.2 EUR InStock
PRTWFT15BKB36 PRTWFT15BKB37 PRTWFT15BKB38 PRTWFT15BKB39 PRTWFT15BKB40 PRTWFT15BKB41 PRTWFT15BKB42 PRTWFT15BKB43 PRTWFT15BKB44 PRTWFT15BKB45 PRTWFT15BKB46 PRTWFT15BKB47 PRTWFT15BKB48
Steelite™ Tove Trainer S1P Safety Shoes, Lace-up, Low
Schuhe und Stiefel
The Steelite Tove Trainer combines a modern trainer style with classic safety protection.
  • Direct injected mesh with KPU cage, providing strength and breathability
  • Extra set of contrast laces included
  • Protective steel toecap
  • Pierce-resistant steel midsole
  • SRC - slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Anti-static footwear
  • Energy absorbing seat region
  • Fuel- and oil-resistant outsole
  • Dual density sole unit
The TPU moulded upper with mesh backing offers excellent breathability and comfort. Dual density EVA/rubber outsole for a lightweight and long lasting trainer. Superb comfort footbed with steel toecap and steel midsole.
Zertifizierungen: EN ISO 20345: 2011 S1P. AS 2210.3. CE certified.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge