Winter-Sicherheitsstiefel, Compositelite™, Thor, FC12
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FW05BKR46 FW05BKR47 FW05BKR48 FW05BKR44 FW05BKR45 FW05BKR37 FW05BKR38 FW05BKR39 FW05BKR40 FW05BKR41 FW05BKR42 FW05BKR43
PRTWFW05BKR46EA 56.4 EUR InStock
PRTWFW05BKR46 PRTWFW05BKR47 PRTWFW05BKR48 PRTWFW05BKR44 PRTWFW05BKR45 PRTWFW05BKR37 PRTWFW05BKR38 PRTWFW05BKR39 PRTWFW05BKR40 PRTWFW05BKR41 PRTWFW05BKR42 PRTWFW05BKR43
Winter-Sicherheitsstiefel, Compositelite™, Thor, FC12
Schuhe und Stiefel
Anti-static, high leg winter boot made from water resistant split leather and a durable PU/rubber sole, resistant to oil and heat (200 °C).
  • Fur lining
  • Shock-absorbent heel
  • Protection toecap made from steel (200 J)
  • Puncture resistant midsole
Suitable for use in cold working conditions.
Colour: Black
Zertifizierungen: EN 20345:2011+A1:2007; S3, CI, HRO, SRC.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge