Occupational shoes, slip-on, FW58
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
FW58WHR48 FW58WHR34 FW58WHR37 FW58WHR42 FW58WHR41 FW58WHR35 FW58WHR44 FW58WHR36 FW58WHR40 FW58WHR38 FW58WHR39 FW58WHR43 FW58WHR47 FW58WHR45 FW58WHR46
PRTWFW58WHR48EA 45.5 EUR InStock
PRTWFW58WHR48 PRTWFW58WHR34 PRTWFW58WHR37 PRTWFW58WHR42 PRTWFW58WHR41 PRTWFW58WHR35 PRTWFW58WHR44 PRTWFW58WHR36 PRTWFW58WHR40 PRTWFW58WHR38 PRTWFW58WHR39 PRTWFW58WHR43 PRTWFW58WHR47 PRTWFW58WHR45 PRTWFW58WHR46
Occupational shoes, slip-on, FW58
Schuhe und Stiefel
Washable upper with a self-cleaning outsole makes this slip on shoe ideal when a protective toecap is not required. Lightweight and comfortable construction.
  • Anti-static footwear
  • Energy absorbing seat region
  • Water resistant upper to prevent water penetration
  • Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Fuel and oil resistant outsole
  • Mono density sole unit
Material upper: Microfibre
Material sole: PU
These shoes do not utilise animal product or biproducts.
Zertifizierungen: CE certified, EN ISO 20347 O2
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge