Wrap around glasses, PW32
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PW32CLR
PRTWPW32CLREA 4.7 EUR InStock
PRTWPW32CLR
Wrap around glasses, PW32
Schutzbrillen
Wrap-around design glasses with one piece lens.
  • 99% UV protection
  • Anti-scratch coating for added durability
  • Flexible arms for ultra comfort
  • Ultra lightweight for long uses
  • Individually packed for vending machines
  • Retail bag which aids presentation for retail sales
Zertifizierungen: AS/NZS 1337.1; EN 170 (2C-1.2 (clear)); ANSI/ISEA Z87 + (U6 (Clear))
Lieferumfang: Cord included.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge