Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Holster work shorts, PW345

Lieferant: Portwest Clothing

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

PW345ZBR30 PW345ZBR32 PW345ZBR33 PW345ZBR34 PW345ZBR36 PW345ZBR38 PW345ZBR40 PW345ZBR41 PW345ZBR42
PRTWPW345ZBR30EA 51.3 EUR InStock
PRTWPW345ZBR30 PRTWPW345ZBR32 PRTWPW345ZBR33 PRTWPW345ZBR34 PRTWPW345ZBR36 PRTWPW345ZBR38 PRTWPW345ZBR40 PRTWPW345ZBR41 PRTWPW345ZBR42
Holster work shorts, PW345
Arbeitskleidung
The holster work shorts are a great addition to the PW3 range, combining function and style and impressive range of multifunctional pockets including detachable holster pockets, ID pocket, rule pocket and an easy access thigh pocket provide ample secure storage.

  • Durable polycotton fabric for high performance and maximum wearer comfort
  • Hammer loop for secure storage of work tools
  • Zip off holster pockets for added versatility
  • Double rule pocket
  • Triple-stitched seams for extra durability
  • Reinforced panels in high wear areas for maximum durability

Contrast fabric: 100% polyester oxford 600D 210 g.

Shell fabric: Kingsmill 300 g.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge