Welding safety spectacle, PW68
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PW68BGR
PRTWPW68BGREA 7.5 EUR InStock
PRTWPW68BGR
Welding safety spectacle, PW68
Schutzbrillen
Multipurpose Shade 5 lens with lateral protection and adjustable arms for customised comfort.
  • 99% UV protection
  • Anti-scratch coating for added durability
  • Free spec cord
  • Adjustable length for a secure and comfortable fit
  • Excellent side protection
  • Ultra lightweight for long uses
  • Individually packed for vending machines
  • Retail bag which aids presentation for retail sales
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge