Browguard with visor, PP
Lieferant: Portwest Clothing
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PW91CLR
PRTWPW91CLREA 24.3 EUR InStock
PRTWPW91CLR
Browguard with visor, PP
Gesichtsschutz
PC visor with PP browguard, is manufactured from impact resistant PP.
  • Wheel ratchet adjustment system is designed for easy use
  • Replacement visor available (PW92)
Zertifizierungen: EN 166 (1 B/3 B)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge