Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Hi-vis reversible bodywarmer vests, S469

Lieferant: Portwest Clothing

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

S469ORR5XL S469ORRL S469ORRXL S469ORRXXL S469ORRM S469ORRS S469ORRXXXL S469ORR4XL S469YER4XL S469YERL S469YERM S469YERS S469YERXL S469YERXS S469YERXXL S469YERXXXL S469YER5XL
PRTWS469ORR5XLEA 46.5 EUR InStock
PRTWS469ORR5XL PRTWS469ORRL PRTWS469ORRXL PRTWS469ORRXXL PRTWS469ORRM PRTWS469ORRS PRTWS469ORRXXXL PRTWS469ORR4XL PRTWS469YER4XL PRTWS469YERL PRTWS469YERM PRTWS469YERS PRTWS469YERXL PRTWS469YERXS PRTWS469YERXXL PRTWS469YERXXXL PRTWS469YER5XL
Hi-vis reversible bodywarmer vests, S469
Arbeitskleidung
These multi use high visible bodywarmer vests are specially designed for those who will not compromise on style, comfort, protection and performance. These garments are designed to keep warm, dry and visible in all conditions. The 300D Oxford Industry High Visibility provides comprehensive range of single and combination garments in a highly durable and cleverly designed innovative styles.

  • Taped seams to provide additional protection
  • Reflective tape for increased visibility
  • Quilt lined for thermal insulation
  • Waterproof with sealed seams
  • Zipper front closure
  • 50+ UPF rated fabric to block 98% of UV rays

Shell fabric is made of 100% polyester, 300D Oxford weave with a stain resistant finish, PU coated (190 g), Lining fabric is made of 65% polyester, 35% cotton (260 g) and Filling fabric is made of 100% polyester (200 g).

Features with 2" silver reflective tape to promote wearer safety. Designed with four pockets that is two welt with flaps on outside, two lower patch on reverse side. Extremely water resistant fabric finish, water beads away from fabric surface.

Zertifizierungen: ANSI/ISEA 107: 2015 TYPE R CLASS 2; AS 4602.1 Class D/N; RIS-3279-TOM ISSUE 1 (Orange only; EN ISO 20471 Class 2.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge