Proteomic protein controls
Lieferant: G Bioscienes
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PSC-01 PSC-02 PSC-03 PSC-04 PSC-05
PSC-01EA 299 EUR InStock
PSC-01 PSC-02 PSC-03 PSC-04 PSC-05
Proteomic protein controls
Protein-Aufbereitungs Proteinreinigungs-Reagenzien
Protein preparations specifically prepared for standardising electrophoresis methods and protocols. Substantially free from non-protein agents such as nucleic acids, detergents, salts, lipids and other common laboratory agents, the protein preparation has low conductivity (<50 μs). Supplied as dry protein pellets, simply re-hydrate and use. Proteomic protein sample controls are available for animal cells, E.coli, yeast, plant or as a control set that contains one 2 mg vial of each.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge