Multi layer adhesive floor mats
Lieferant: PURUS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PM264538W-HT PM264538G PM264538W PM184538W PM184538W-HT PM364534B PM364564G PM264538B-HT PM243638W PM264538B
PURUPM264538W-HTEA 354 EUR InStock
PURUPM264538W-HT PURUPM264538G PURUPM264538W PURUPM184538W PURUPM184538W-HT PURUPM364534B PURUPM364564G PURUPM264538B-HT PURUPM243638W PURUPM264538B
Multi layer adhesive floor mats
Fußmatten Klebematten
A choice of either 30 or 60 layer adhesive entrance mats are manufactured from low density polyethylene. The mats control contamination by removing debris from the soles of shoes and the wheels of transport containers. When one layer is dirty, simply remove by the numbered corner tab, leaving a fresh layer ready for use.
  • Antimicrobial agent protects against the growth of bacteria and fungi
  • Non-slip, low profile, individually numbered layers
  • Reduced maintenance and cleaning costs
Colour: Clear, white, grey or blue
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge