Drawer Racks, Stainless Steel
Lieferant: RATIOLAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
5400046 5400019 5400050 5400047
RATI5400046EA 176.3 EUR InStock
RATI5400046 RATI5400019 RATI5400050 RATI5400047
Drawer Racks, Stainless Steel
Gestelle Kryogestelle
Stainless steel racks for storing the cryo boxes in freezer cabinets and chests.
  • Cabinet slide-ins without pull-out stop
  • Easy-to-clean polished surface
  • Robust design
Racks for for all cryo boxes format 130×130; 133×133 and 136×136 mm.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge