Deep well blocks, 96-well, low profile
Lieferant: RATIOLAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
8101012
RATI8101012EA 148.35 EUR InStock
RATI8101012
Deep well blocks, 96-well, low profile
Mikrotiterplatten Deep-Well-Mikroplatten
These 1.2 ml low-profile deep-well plates made from clear PP are ideal for pharmaceutics, biotechnology and diagnostics.
  • Well with significantly larger cross section and raised rims for reduced cross-contamination
  • Autoclavable at 121 °C, 20 minutes
  • Temperature resistant to –80 °C
  • 8 x 12 wells with alphanumeric code in micro test plate format
  • Precise design, for use in automatic pipetting systems for High-Throughput-Screening (HTS)
  • Optimum space for bar code labels on the side walls
  • Space-saving and stackable
Being of the same standard dimensions (Society for Biomolecular Sciences – SBS), they are one-third lower than standard deep well plates (96). Therefore, they require less space. Requiring around 30% less space, they help to optimize warehouse space, hence lowering costs – especially for costly cryo-storage applications.
The cavities are 20% larger than conventional plates. Thus, when used with robots, they provide additional security. The chimney form of the wells reliably prevents cross-contamination. Their precise, true-to-size form makes them highly suitable for High-Throughput-Screening (HTS).
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge