RIDASCREEN®FAST Enzyme assay, lysozym EIA
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
R6452
RBIOR6452EA 670 EUR InStock
RBIOR6452
RIDASCREEN®FAST Enzyme assay, lysozym EIA
Tests Immunologische Tests
RIDASCREEN®FAST Lysozym is a sandwich enzyme immunoassay for the quantitative analysis of lysozyme (hen’s egg protein) in foods like wine, cheese and sausage.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge