Carbon standards
Lieferant: REAGECON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
TOIC10 TOIC100 TOIC1M TOC50 TIC1M TOC1M TIC500 TOC500 TOC2M TOC5 TIC5 TOC100 TOC10 TOC20 TOC15 TOC250
REGETOIC10EA 369 EUR InStock
REGETOIC10 REGETOIC100 701-0049 REGETOC50 REGETIC1M REGETOC1M REGETIC500 REGETOC500 REGETOC2M REGETOC5 REGETIC5 REGETOC100 REGETOC10 REGETOC20 REGETOC15 REGETOC250
Carbon standards
Standards Kationischer Standards Kohlenstoff-Standards
These standards are manufactured from high purity raw materials and is suitable for calibrating a wide range of Total Organic Carbon (TOC) and Total Inorganic Carbon (TIC) analysers to a high accuracy.
These standards are independent, traceable and certified.
Zertifizierungen: Accredited to IEC/ISO 17025 and ISO 17034.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge