Column kit, Stx-CLPesticides
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
11190
REST11190EA 1648 EUR InStock
REST11190
Column kit, Stx-CLPesticides
Chromatographiesäulen
Application specific columns for organochlorine pesticides and herbicides.
  • Baseline separations in less than 10 minutes
  • Temperature range: stable to 330 °C
  • Siltek surface deactivation enhances responses for endrin, DDT, methoxychlor
Siltek passivation technology enables these columns to offer unsurpassed inertness and the highest responses for active pesticides.
Lieferumfang: Kit contains an Stx-CLPesticides column, Stx-CLPesticides2 column, Universal angled 'Y' press-tight connector, and a Siltek-deactivated guard column. Columns are not preconnected in this kit.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge