Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

PLOT columns, Rt-Q-BOND columns (fused silica)

Lieferant: Restek

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

19744 19742 19765 19741
REST19744EA 857 EUR InStock
REST19744 REST19742 REST19765 REST19741
PLOT columns, Rt-Q-BOND columns (fused silica)
Chromatographiesäulen
Rt®-Q-BOND are nonpolar PLOT columns incorporating 100% divinylbenzene.

  • Analysis of C1 to C3 hydrocarbons as well as isomers and alkanes up to C12
  • Use for analysis of oxygenated compounds and solvents
  • Maximum temperature of 300 °C

High retention for CO₂ simplifies gas analysis. CO₂ and methane separated from O₂/N₂/CO (Note: O₂/N₂/CO not separated at ambient temperature)

Similar phase(s): HP PLOT Q, CP-PoraPLOT Q, CP-PoraBOND Q, Supel-Q-PLOT, AT-Q.

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge