Open end wrench set
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
20387
REST20387EA 155 EUR InStock
REST20387
Open end wrench set
Schraubenschlüssel
High-quality 1/4×5/16", 3/8"×7/16", 7/16"×1/2", and 1/2"×9/16" wrenches for tightening a wide variety of chromatography fittings.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge