Septum nuts
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
21309 20631
REST21309EA 72 EUR InStock
REST21309 REST20631
Septum nuts
Fittings und Verbinder
These septum nuts are made of high-quality stainless steel and for use with Agilent 5890/6850/6890/7890 split or splitless injectors.
  • Needle guide allows easy penetration and prevents septum coring
  • Manual injection septum nut allows use of 26-gauge needles for on-column injections
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge