ECD vent trap
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
22017
REST22017EA 90 EUR InStock
REST22017
ECD vent trap
Filter Gasfilter
This trap reduces the release of hazardous materials from the ECD vent into the lab.
Dimensions: 152×25 mm
Lieferumfang: Includes connecting lines and mounting kit.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge