Mini regulator
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
22032
REST22032EA 611 EUR InStock
REST22032
Mini regulator
Druckminderer
This mini regulator is suitable for natural gas and refinery gas standards.
Inlet pressure range: 0 to 20,68 bar
Outlet pressure range: 0 to 1,03 bar
Lieferumfang: Supplied with outlet pressure gauge (0 - 1,03 bar), brass CGA 170 nut and nipple, and compression outlet (3,175 mm).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge