Capillary installation gauges
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
21034 22330 22335
REST21034EA 0 EUR InStock
REST21034 REST22330 REST22335
Capillary installation gauges
Werkzeug zum Herausziehen und Einführen
These capillary installation gauges for Agilent GCs are made from high-quality stainless steel.
  • Prevents crushed column ends
  • Seats graphite ferrules onto column for consistent installations
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge