Gas-tight syringes, PTFE tip (SGE)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
24725 24726 24727 24728 24729 24730 24734 24735 24737 24738 24739 24741 24742 24743 24750 24752 24755 24757 24759 24760 24761 24762
REST24725EA 94.4 EUR InStock
REST24725 REST24726 REST24727 REST24728 REST24729 REST24730 REST24734 REST24735 REST24737 REST24738 REST24739 REST24741 REST24742 REST24743 REST24750 REST24752 REST24755 REST24757 REST24759 REST24760 REST24761 REST24762
Gas-tight syringes, PTFE tip (SGE)
Spritzen Spritzen für die Chromatographie
These gas-tight syringes with interchangeable barrels and plungers and tips extend performance and increase cost effectiveness.
  • Suitable for gases and liquids
  • High accuracy of dispensed volumes
Bestellinformation: Needles have to be ordered separately for syringes without needles.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge