Bulk adsorbents, Florisil® PR
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
26135
REST26135EA 335 EUR InStock
REST26135
Bulk adsorbents, Florisil® PR
Chromatographie-Sorbentien SPE-Sorbentien
Florisil® PR is commonly used to remove polar interferences from pesticide residues. This bulk material is ideal for labs packing their own extraction cartridges for pesticide residue extractions.
  • Pesticide residue grade
Verpackung: Packaged in glass containers.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge