HPLC performance kit
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
26394
REST26394EA 268 EUR InStock
REST26394
HPLC performance kit
Chromatographie-Zubehör Chromatographie-Ersatzteile
This HPLC PEEK performance kit is a useful complement for the HPLC system.
Lieferumfang: It includes 1 hub-cap adapter w/cap, 3 tubing clips, 1 clean cut tubing cutter, 5 PEEK finger tight fittings, 5 PEEK column plugs, 2 PEEK 90° tubing elbows, 2 PEEK 180° tubing elbows and 2 PEEK column connectors.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge