Trident LC Column Protection System
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
27473
REST27473EA 487 EUR InStock
REST27473
Trident LC Column Protection System
Chromatographiesäulen
Trident LC column protection system is available in three levels of protection such as filter only, cartridge only, or filter and cartridge.
  • Easy-to-remove cap frit simplifies filter replacement
  • Low-dead-volume design has negligible effect on chromatography
  • Direct connection eliminates tubing and connectors that increase system volume and leak potential
The durable metal tip with replaceable PEEK ferrule means easy installation onto column without tools. Improved thread design and materials create an optimal seal that releases and reseals easily, allowing multiple installations without galling and binding.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge