524 Calibration mix #8 (12 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30203
REST30203EA 80.5 EUR InStock
REST30203
524 Calibration mix #8 (12 components)
Standards Organische Standards
Mix Contains: Carbon disulphide (75-15-0); Chloroacetonitrile (107-14-2); 1-Chlorobutane (Butyl chloride) (109-69-3); trans-1,4-Dichloro-2-butene (110-57-6); 1,1-Dichloro-2-propanone (513-88-2); Diethyl ether (ethyl ether) (60-29-7); Hexachloroethane (67-72-1); Iodomethane (methyl iodide) (74-88-4); Methacrylonitrile (126-98-7); Methyl tert-butyl ether (MTBE) (1634-04-4); 2-Nitropropane (79-46-9); Propionitrile (107-12-0).
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge