8240 Alcohols mix (5 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30214
REST30214EA 57 EUR InStock
REST30214
8240 Alcohols mix (5 components)
Standards Organische Standards
Mix contains: Allyl alcohol (2-propen-1-ol) (107-18-6); 2-Chloroethanol (107-07-3); Ethanol (64-17-5); Isobutyl alcohol (78-83-1); Propargyl alcohol (107-19-7).
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge