8240 Nitriles mix (7 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30215
REST30215EA 69.5 EUR InStock
REST30215
8240 Nitriles mix (7 components)
Standards Organische Standards
Mix contains: Acrylonitrile (107-13-1); Ethyl methacrylate (97-63-2); Malononitrile (109-77-3); Methacrylonitrile (126-98-7); Methyl methacrylate (80-62-6); Propionitrile (107-12-0); Styrene (100-42-5).
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge