Certified BTEX in unleaded gas composite standard (9 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30237
REST30237EA 62 EUR InStock
REST30237
Certified BTEX in unleaded gas composite standard (9 components)
Standards Organische Standards
The BTEX in unleaded gas composite standard consists of 9 components, they are benzene, ethylbenzene, isopropyl benzene, methyl tert-butyl ether (MTBE), naphthalene, toluene, m-xylene, o-xylene and p-xylene.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge