MA VPH standard with surrogate
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30452
REST30452EA 77 EUR InStock
REST30452
MA VPH standard with surrogate
Standards Mineralölindustrie-Standards
The MA VPH standard consists of 14 components they are (C5) n-pentane 1000 μg/ml, (C9) n-nonane 1000 μg/ml, benzene 500 μg/ml, 2,5-dibromotoluene (SS) 1000 μg/ml, ethylbenzene 500 μg/ml, 2-methylpentane 1500 μg/ml, methyl tert-butyl ether (MTBE) 1500 μg/ml, naphthalene 1000 μg/ml, toluene 1500 μg/ml, 1,2,4-trimethylbenzene 1000 μg/ml, 2,2,4-trimethylpentane (isooctane) 1500 μg/ml, m-xylene 1,000 μg/ml, o-xylene 1,000 μg/ml and p-xylene 1,000 μg/ml.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge