MA VPH matrix spike mix with surrogate (14 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30454
REST30454EA 79.5 EUR InStock
REST30454
MA VPH matrix spike mix with surrogate (14 components)
Standards Mineralölindustrie-Standards
The MA VPH matrix spike mix consists of 14 components they are (C5) n-pentane, (C9) n-nonane, benzene, 2,5-dibromotoluene (SS), ethylbenzene, 2-methylpentane, methyl tert-butyl ether (mtbe), naphthalene, toluene, 1,2,4-trimethylbenzene, 2,2,4-trimethylpentane (isooctane), m-xylene, o-xylene and p-xylene.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge