Certified aromatics in gasoline, 16 components
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30485
REST30485EA 134 EUR InStock
REST30485
Certified aromatics in gasoline, 16 components
Standards Mineralölindustrie-Standards
Confirms unleaded gasoline TPH, BTEX and aromatics in single analysis.
This standard consists of 16 components.
Components: Benzene, ethylbenzene, m-ethyltoluene, o-ethyltoluene, p-ethyltoluene, isopropylbenzene, methyl tert-butyl ether (MTBE), naphthalene, n-propylbenzene, toluene, 1,2,3-trimethylbenzene, 1,2,4-trimethylbenzene, 1,3,5-trimethylbenzene, m-xylene, o-xylene, p-xylene.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge