NJDEP EPH 10/08 rev. 2 aliphatics matrix spike mix (18 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30542
REST30542EA 135.5 EUR InStock
REST30542
NJDEP EPH 10/08 rev. 2 aliphatics matrix spike mix (18 components)
Standards Organische Standards
Consists of 18 components, those are n-nonane, n-decane,  n-dodecane,  n-tetradecane, n-hexadecane,  n-octadecane,  n-eicosane,  n-heneicosane,  n-docosane,  n-tetracosane,  n-hexacosane,  n-octacosane,  n-triacontane,  n-dotriacontane,  n-tetratriacontane,  n-hexatriacontane, n-octatriacontane and n-tetracontane.
18 components
Zertifizierungen: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge