Ethylene oxide standard
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30548
REST30548EA 113.5 EUR InStock
REST30548
Ethylene oxide standard
Standards Organische Standards
Used as reference standards for detecting ethylene oxide and 1,4-dioxane in PEG-based products.
  • Formulated with concentrations needed by labs analysing cosmetics and personal care products for possible carcinogens
  • Quantitatively tested to confirm composition; detailed support documentation provided
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge