Oxygenates (6 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
30626
REST30626EA 71.5 EUR InStock
REST30626
Oxygenates (6 components)
Standards Organische Standards
This Oxygenates compound includes six components: tert-amyl ethyl ether (TAEE) 2000 μg/ml, tert-Amyl methyl ether (TAME) 2000 μg/ml, tert-Butanol (TBA) 10000 μg/ml, Diisopropyl ether (DIPE) 2000 μg/ml, Ethyl-tert-butyl ether (ETBE) 2000 μg/ml and Methyl tert-butyl ether (MTBE) 2000 μg/ml.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge