Steroids and mixed pharmaceuticals mix (10 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31117
REST31117EA 85.5 EUR InStock
REST31117
Steroids and mixed pharmaceuticals mix (10 components)
Standards Umweltanalyse-Standards
This steroids and mixed pharmaceuticals mix contain ten components: Bisphenol A, Diclofenac sodium salt, 17-beta-Estradiol, Estrone, 17-alpha-Ethynylestradiol, 4-para-Nonylphenol, 4-tert-Octylphenol, Primidone, Progestrone and Testosterone.
Shelf life 24 months.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge