QuEChERS performance standard A (16 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31153
REST31153EA 145.5 EUR InStock
REST31153
QuEChERS performance standard A (16 components)
Standards Lebensmittel-Standards
QuEChERS performance standard A designed for use in all QuEChERS methods for pesticides in fruits and vegetables. Ideal for initial method evaluations and ongoing method performance validations.
  • Optimized blend of chemically compatible analytes for maximum stability and shelf life
  • Quantitatively analyzed to confirm the composition and stability of each mixture
Precise formulation improves data quality and operational efficiency; spend more time running samples and less time sourcing and preparing standards.
This standard contain 16 components: Acephate, Azinphos methyl, Chlorpyrifos, Coumaphos, Diazinon, Dichlofluanid, Dichlorvos, Dimethoate, Fenthion, Malathion, Methamidophos, Mevinphos, Omethoate, Phosalone, Pirimiphos methyl and Propargite.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements. It including the original unbuffered method, AOAC 2007.01, and EN15662.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge