Fuel oil degradation mix (4 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31240
REST31240EA 63.5 EUR InStock
REST31240
Fuel oil degradation mix (4 components)
Standards Mineralölindustrie-Standards
Subsurface degradation of fuel oil spills can be estimated by examining the ratios of C17/pristane and C18/phytane. To assist in identifying these four compounds from the complex fuel oil analysis, Restek offer a product that contains these compounds for retention time determination.
Fuel oil degradation mix contains (C17) heptadecane, (C18) octadecane, pristane (2,6,10,14-tetramethylpentadecane), phytane (2,6,10,14-tetramethylhexadecane).
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge