8330 calibration mix #2 (7 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31451
REST31451EA 82.5 EUR InStock
REST31451
8330 calibration mix #2 (7 components)
Standards Umweltanalyse-Standards
EPA method 8330 is used to measure explosives residues in water and soil samples, using HPLC with UV detection. Target analytes are nitroaromatic and nitramine explosives and their degradation products. Obtaining pure, neat compounds for Method 8330 reference standards can be very difficult. To ensure the highest quality standards, Restek's Quality Control (QC) lab confirms the identity and purity of mixture components and solvents using one or more of the following techniques: GC-FID, HPLC, GC-ECD, GC-MS, LC-MS, refractive index, and melting point.
8330 calibration mix #2 contains 2-amino-4,6-dinitrotoluene, 4-amino-2,6-dinitrotoluene, 2,6-dinitrotoluene, 2-nitrotoluene, 3-nitrotoluene, 4-nitrotoluene, tetryl.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge