610 PAH calibration mix B (16 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31455
REST31455EA 97 EUR InStock
REST31455
610 PAH calibration mix B (16 components)
Standards Umweltanalyse-Standards
This 610 PAH calibration mix B idela for LC-UV detection.
It consists of 16 components: Acenaphthene (1000 μg/ml), Acenaphthylene (2000 μg/ml), Anthracene(100 μg/ml), Benz(a)anthracene (100 μg/ml), Benzo(a)pyrene (100 μg/ml), Benzo(b)fluoranthene (200 μg/ml), Benzo(k)fluoranthene (100 μg/ml), Benzo(ghi)perylene (200 μg/ml), Chrysene (100 μg/ml), Dibenz(a,h)anthracene (200 μg/ml), Fluoranthene (200 μg/ml), Fluorene (200 μg/ml), Indeno(1,2,3-cd)pyrene (100 μg/ml), Naphthalene (1000 μg/ml), Phenanthrene (100 μg/ml) and Pyrene (100 μg/ml).
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge