MA EPH matrix spike mix (10 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31460
REST31460EA 63 EUR InStock
REST31460
MA EPH matrix spike mix (10 components)
Standards Umweltanalyse-Standards
Contains: n-nonane, C9, n-tetradecane, C14, n-nonadecane, C19, n-eicosane, C20, n-octacosane, C28, acenaphthene, anthracene, chrysene, naphthalene , pyrene.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge