8095 Matrix spike mix B (7 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31610
REST31610EA 49 EUR InStock
REST31610
8095 Matrix spike mix B (7 components)
Standards Organische Standards
The mix consists of, 3,5-dinitroaniline, nitrobenzene, nitroglycerin, 2-nitrotoluene, 3-nitrotoluene, 4-nitrotoluene, PETN.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge