Calibration mixture, Connecticut ETPH
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31614
REST31614EA 79.5 EUR InStock
REST31614
Calibration mixture, Connecticut ETPH
Standards Gaschromatographie-Standards
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs.
  • 15 components- all n- alkanes
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge