Standard mixture of n-alkanes for system performance test
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31633
REST31633EA 108.5 EUR InStock
REST31633
Standard mixture of n-alkanes for system performance test
Standards Organische Standards
These are reference mixtures for ISO/DIS 9377 (German H-53), a gas chromatography–flame ionization detection (GC-FID) method.
  • For GC analysis of total petroleum hydrocarbons (TPH) in water
  • Environmentally safer than previous methods
  • Calibration standard available as diesel no.2/motor oil and diesel no.2/mineral oil
Concentration: 50 μg/ml each in cyclohexane.
Zertifizierungen: ISO/DIS 9377 Water Quality Testing (German H-53)
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge