Haloacetic acid methyl ester mix #1 (6 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
31645
REST31645EA 126 EUR InStock
REST31645
Haloacetic acid methyl ester mix #1 (6 components)
Standards Organische Standards
The mix consists of methyl bromochloroacetate, methyl dibromoacetate, methyl dichloroacetate, methyl monobromoacetate, methyl monochloroacetate, methyl trichloroacetate.
  • Prepared from purified neat reference materials for the highest quality
  • Lot-to-lot consistency ensures analytical reproducibility
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements. Meet Stage 1 requirements for US EPA Methods 552, 552.1 and 552.2 and meet requirements for Stage 2 DBPR.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge